Lister Bandage Scissors
Supplier: WORLD PRECISION INSTRUMENTS LLC
500444 500444-G 501701 501702
77662-342EA 46.22 USD
77662-342 77662-344 77662-660 77662-662
Lister Bandage Scissors
Scissors
Lister bandage scissors, also called Lister scissors or Lister dressing scissors, are a pair of surgical scissors designed for cutting and removing dressings, bandages, and medical materials without hurting the patient.
  • Blunt, angled tips
  • With 4 cm cutting edge
  • Autoclavable
  • Surgical grade stainless steel or premium German steel
The blunt tip prevents accidental injury or puncturing of the patient's skin while cutting close to the body. The angled design lets you easily insert the scissors under bandages. The angled blade also gives you better visibility and improved access to the dressing, especially in tight or confined spaces. The cutting blade is blunt.
These surgical scissors are primarily used for removing dressings, bandages, or gauze. This is common in wound care, postoperative care, and first aid.