MiniStar™ Miniature Peristaltic Pumps
Supplier: WORLD PRECISION INSTRUMENTS LLC
504011 MINISTAR
77663-432EA 1373.56 USD
77663-432 77664-822
MiniStar™ Miniature Peristaltic Pumps
Pumps Peristaltic Pumps
This compact, lightweight peristaltic pump fits just about anywhere. It can be mounted directly on the bench, in a regular rack or to a post.
  • Remote control (wired)
  • Small footprint
  • Quiet operation
  • Compact and easily transported
The speed can be adjusted from 1 to 50 rpm. With recommended silicone tubing, the volume can be set from 0.06 to 14.0 ml/min. The MiniStar™ also features a hand held remote control that allows you to start and stop the pump, purge or adjust its speed and direction.
Applications are small volume chemical transfer, pharmaceutical processing and sample perfusion.
Información para pedidos: MiniStar is sold separately with out the stand and clamp shown in the image, 504011 system includes the MiniStar, stand and clamp.
Información de suministro: Includes tubing.