Human Recombinant IL-15 (CTGrade GMP) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
500-08-100UG 500-08-1MG 500-08-50UG
77660-986EA 1444.06 USD
77660-986 77660-988 77660-990
Human Recombinant IL-15 (CTGrade GMP) (from E. coli)
Proteins and Peptides
IL-15 is a widely expressed proinflammatory cytokine that is structurally and functionally related to IL-2.
  • High quality
  • Low endotoxin
  • Lot-to-lot consistency
  • Animal free
  • Manufactured following GMP guidelines
Signals through JAK kinases to activate STAT3, STAT5, and STAT6 transcription factors
Regulates the activation of T cells, neutrophils, and macrophages, and is a stimulatory cytokine promoting dendritic cell function
Dysregulated in chronic inflammatory disorders, including rheumatoid arthritis, psoriasis, and pulmonary inflammatory diseases
Combines with other common γ chain cytokines (IL-2, IL-7 and IL-21) to promote favorable anti-tumoral phenotypes in adoptive cellular therapies such as CAR-T cell therapy
The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance
Host Cell Protein: ≤30 ng/ml
Host Cell DNA: ≤0.01 ng/ml
Sterility testing: No Growth
Mycoplasma: Negative
Precaución: For research use or further manufacturing only.