Petri Dishes 92×16 mm

Supplier: SARSTEDT INC
82.1195 82.1473 82.1473.001 82.1473.020 82.1473.040 82.1473.060 82.1473.080
77589-164CS 233.22 USD
77589-164 77589-168 77589-170 77589-172 77589-174 77589-176 77589-178
Petri Dishes 92×16 mm
Culture Dishes Petri Dishes
One aspect of microbiology is researching and identifying pathogens, as well as looking for suitable treatments. SARSTEDT provides researchers with the consumables they need to do their work.

  • Ø×height: 92×16 mm
  • Ventilation cams: Yes
  • Ideal for cultivating microorganisms on and in solid culture media
  • Petri dishes with 3 ventilation cams in the lid for optimal gas exchange and reduced condensation
  • Fully automated production from crystal-clear polystyrene
  • Resistant to applications with approx. 60 °C hot and liquid agar
  • Can be easily stacked and used in common automated plate pouring systems

For the cultivation of microorganisms, SARSTEDT offers Petri dishes made of high-quality, crystal-clear polystyrene. Due to their high dimensional stability, they are ideal for use in automatic plate filling systems and are easily stacked.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR