Cell Culture Plates, 6-well

Supplier: SARSTEDT INC
83.3920 83.3920.005 83.3920.300 83.3920.500
77589-300CS 163.58 USD
77589-300 77589-302 77589-304 77589-306
Cell Culture Plates, 6-well
Microplates Cell Culture Plates
SARSTEDT offers a wide range of flasks, dishes and plates made of high-quality, transparent polystyrene. Many formats are available with all three growth surfaces. Every container can be easily identified by SARSTEDT’s color-guided system, the batch number and the expiration date even after it has been removed from the packaging.

  • Growth area per well: 9.07 cm²
  • Material: PS
  • Number of wells: 6
  • Base shape: Flat base
  • Working volume: 4 ml
  • Complies with ANSI/SLAS standards, ensuring compatibility with many automated laboratory instruments
  • Available with three different growth surfaces, easy to identify thanks to clear color markings
  • Free standing wells with alphanumeric labels minimize the risk of contamination during pipetting
  • Condensation rings and ventilation cams in the lid ensure the constant exchange of gas while minimizing evaporation
  • For better traceability, each plate is labeled with a batch number and expiration date

Adherent cells have a wide variety of demands on their growth surface. While most adherent cell lines can be cultivated on the red standard surface, primary cells, sensitive cell culture lines and cells that are cultivated under serum-reduced or serum-free conditions are particularly challenging and need more support to grow successfully on the surface. In these cases, the growth surface often has to be coated with proteins (e.g. polylysine). The properties of the SARSTEDT Cell+ surface (yellow) can eliminate the need for self-coated culture vessels in many cases, making cultivation easier and yielding more reproducible results. A special treatment of the plastic surface introduces additional polar groups into the hydrophilic surface, so that challenging adherent cells are able to adhere and grow without an additional protein coating.

The green surface is ideal for cultivating suspension cells. This hydrophobic surface minimizes cell losses during subcultivation caused by the unwanted microadhesion of cells.

Purity: TC Tested
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR