Cell Culture Flasks, T-25

Supplier: SARSTEDT INC
83.3910 83.3910.002 83.3910.300 83.3910.302 83.3910.500 83.3910.502
77589-264CS 420.38 USD
77589-264 77589-266 77589-268 77589-270 77589-272 77589-274
Cell Culture Flasks, T-25
Flasks Cell Culture Flasks
SARSTEDT offers a wide range of flasks, dishes and plates made of high-quality, transparent polystyrene. Many formats are available with all three growth surfaces. Every container can be easily identified by SARSTEDT’s color-guided system, the batch number and the expiration date even after it has been removed from the packaging.

  • Growth area: 25 cm²
  • Material: PS
  • Cap material: PE
  • Working volume: 7 ml to 12.5 ml
  • With canted neck to prevent overflowing of the medium and associated contamination; facilitates work through
  • fields for labeling and high traceability with lot no. and expiration date on each bottle
  • Bottle geometry allows all corners to be reached with serological pipettes or cell scrapers
  • Quick-closure cap - in the filter cap and 2-position screw cap versions - allows opening and closing
  • with 1/3 rotation
  • Available with three different growth surfaces, clear identification through the colored lid
  • Packed in a resealable minigrip bag, sealed with a tamper-evident closure before opening for the first time

Adherent cells have a wide variety of demands on their growth surface. While most adherent cell lines can be cultivated on the red standard surface, primary cells, sensitive cell culture lines and cells that are cultivated under serum-reduced or serum-free conditions are particularly challenging and need more support to grow successfully on the surface. In these cases, the growth surface often has to be coated with proteins (e.g. polylysine). The properties of the SARSTEDT Cell+ surface (yellow) can eliminate the need for self-coated culture vessels in many cases, making cultivation easier and yielding more reproducible results. A special treatment of the plastic surface introduces additional polar groups into the hydrophilic surface, so that challenging adherent cells are able to adhere and grow without an additional protein coating.

The green surface is ideal for cultivating suspension cells. This hydrophobic surface minimizes cell losses during subcultivation caused by the unwanted microadhesion of cells.

Purity: TC Tested
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR