1-(5-Bromo-2-fluorophenyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

99816-5G 99816-25G 99816-1G 99816-500MG
77630-950EA 1441 USD
77630-950 77660-604 77644-266 77652-264
1-(5-Bromo-2-fluorophenyl)-1H-pyrazole ≥97%
1-(5-Bromo-2-fluorophenyl)-1H-pyrazole

Precaución: For Research Use Only

Formula: C₉H₆BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD34776081
CAS Number: 425379-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR