4-(3,5-Dimethylphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

99468-5G 99468-1G 99468-25G 99468-500MG
77633-896EA 1206 USD
77633-896 77640-266 77657-132 77648-264
4-(3,5-Dimethylphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3,5-Dimethylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 206.281 g/mol
MW: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD11849470
CAS Number: 499157-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR