4-(4-(Tert-butyl)phenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

99417-5G 99417-25G 99417-1G 99417-500MG
77633-894EA 1381 USD
77633-894 77660-602 77644-264 77652-262
4-(4-(Tert-butyl)phenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(4-(Tert-butyl)phenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 234.334 g/mol
MW: C₁₅H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD19120976
CAS Number: 853886-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR