4-(3-(Benzyloxy)phenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

99400-5G 99400-1G 99400-500MG
77630-948EA 1508 USD
77630-948 77644-262 77652-260
4-(3-(Benzyloxy)phenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3-(Benzyloxy)phenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 284.35 g/mol
MW: C₁₈H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD34776005
CAS Number: 133739-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR