4-(3-Isopropylphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

99386-5G 99386-1G 99386-500MG
77657-130EA 1898 USD
77657-130 77644-260 77652-258
4-(3-Isopropylphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3-Isopropylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 220.307 g/mol
MW: C₁₄H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD31694051
CAS Number: 2027667-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR