4-(3-Chlorophenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

99383-5G 99383-25G 99383-1G 99383-500MG
77633-892EA 1381 USD
77633-892 77657-128 77648-260 77648-262
4-(3-Chlorophenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3-Chlorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 212.673 g/mol
MW: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD18296217
CAS Number: 1339135-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR