4-(3-Methoxyphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

99379-5G 99379-1G 99379-25G 99379-500MG
77633-890EA 1206 USD
77633-890 77640-264 77660-600 77648-258
4-(3-Methoxyphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3-Methoxyphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 208.254 g/mol
MW: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD18296171
CAS Number: 1344057-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR