4-(M-tolyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

99377-5G 99377-25G 99377-1G 99377-500MG
77633-888EA 1206 USD
77633-888 77657-126 77644-258 77648-256
4-(M-tolyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(M-tolyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 192.254 g/mol
MW: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD18296179
CAS Number: 145127-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR