4-Bromo-7-fluorobenzo[b]thiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

99089-5G 99089-25G 99089-1G 99089-500MG
77633-882EA 1221 USD
77633-882 77660-598 77644-256 77648-252
4-Bromo-7-fluorobenzo[b]thiophene-2-carboxylic acid ≥97%
4-Bromo-7-fluorobenzo[b]thiophene-2-carboxylic acid

Precaución: For Research Use Only

Formula: C₉H₄BrFO₂S
Storage Temperature: Ambient
MDL Number: MFCD13180864
CAS Number: 360576-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR