Methyl 4-bromo-7-fluorobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

99088-5G 99088-1G 99088-25G 99088-500MG
77633-880EA 696 USD
77633-880 77640-258 77660-596 77652-254
Methyl 4-bromo-7-fluorobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 4-bromo-7-fluorobenzo[b]thiophene-2-carboxylate

Precaución: For Research Use Only

Formula: C₁₀H₆BrFO₂S
MW: 289.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13180865
CAS Number: 360576-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR