Methyl 5,7-difluorobenzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

99032-5G 99032-25G 99032-1G 99032-500MG
77633-878EA 837 USD
77633-878 77657-120 77644-254 77648-250
Methyl 5,7-difluorobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 5,7-difluorobenzo[b]thiophene-2-carboxylate

Precaución: For Research Use Only

Formula: C₁₀H₆F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD11044565
CAS Number: 550998-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR