3-Bromo-2-fluoro-6-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

98825-5G 98825-1G 98825-500MG
77630-946EA 1719 USD
77630-946 77640-256 77648-246
3-Bromo-2-fluoro-6-(methylthio)pyridine ≥95%
3-Bromo-2-fluoro-6-(methylthio)pyridine

Precaución: For Research Use Only

Formula: C₆H₅BrFNS
MW: 222.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34775768
CAS Number: 2137928-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR