2-Bromo-4,5-dimethoxyaniline ≥97%

Supplier: AOBChem USA

Warning

98808-5G 98808-25G 98808-1G 98808-500MG
77633-874EA 996 USD
77633-874 77657-118 77648-242 77648-244
2-Bromo-4,5-dimethoxyaniline ≥97%
2-Bromo-4,5-dimethoxyaniline

Precaución: For Research Use Only

Formula: C₈H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD08752508
CAS Number: 16791-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR