1-(3-Bromo-2,5-dichlorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

98561-5G 98561-25G 98561-1G 98561-500MG
77630-942EA 1441 USD
77630-942 77657-116 77644-250 77648-240
1-(3-Bromo-2,5-dichlorophenyl)cyclopentanol ≥95%
1-(3-Bromo-2,5-dichlorophenyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₁H₁₁BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD34819573
CAS Number: 2969463-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR