2-Fluoro-3-isopropoxy-4-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

97953-5G 97953-25G 97953-1G 97953-500MG
77630-938EA 1441 USD
77630-938 77657-112 77644-246 77652-248
2-Fluoro-3-isopropoxy-4-methylpyridine ≥95%
2-Fluoro-3-isopropoxy-4-methylpyridine

Precaución: For Research Use Only

Formula: 169.196 g/mol
MW: C₉H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD34775471
CAS Number: 2918931-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR