4-Chloro-5-fluoro-2-methoxyaniline ≥97%

Supplier: AOBChem USA

Warning

95068-5G 95068-25G 95068-1G 95068-500MG
77633-870EA 899 USD
77633-870 77660-588 77644-240 77652-246
4-Chloro-5-fluoro-2-methoxyaniline ≥97%
4-Chloro-5-fluoro-2-methoxyaniline

Precaución: For Research Use Only

Formula: C₇H₇ClFNO
MW: 175.5880 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31380925
CAS Number: 1268392-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR