6-(Cyclopropylmethoxy)-4-methylnicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

94926-5G 94926-25G 94926-1G 94926-500MG
77633-866EA 1320 USD
77633-866 77657-100 77644-236 77648-232
6-(Cyclopropylmethoxy)-4-methylnicotinaldehyde ≥95%
6-(Cyclopropylmethoxy)-4-methylnicotinaldehyde

Precaución: For Research Use Only

Formula: C₁₁H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD22576149
CAS Number: 1289042-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR