1-(4-Fluoronaphthalen-1-yl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

94755-5G 94755-25G 94755-1G 94755-500MG
77630-932EA 1441 USD
77630-932 77657-098 77644-234 77652-244
1-(4-Fluoronaphthalen-1-yl)cyclopentanol ≥95%
1-(4-Fluoronaphthalen-1-yl)cyclopentanol

Precaución: For Research Use Only

Formula: 230.277 g/mol
MW: C₁₅H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD34774244
CAS Number: 2918848-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR