1-(5-Chloro-2-fluoro-4-methylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

94729-5G 94729-25G 94729-1G 94729-500MG
77630-930EA 1441 USD
77630-930 77660-586 77644-232 77652-242
1-(5-Chloro-2-fluoro-4-methylphenyl)cyclopentanol ≥95%
1-(5-Chloro-2-fluoro-4-methylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 228.69 g/mol
MW: C₁₂H₁₄ClFO
Storage Temperature: Ambient
MDL Number: MFCD34774219
CAS Number: 2918829-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR