1-(3,4-Difluoro-2-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

94724-5G 94724-1G 94724-25G 94724-500MG
77630-928EA 1441 USD
77630-928 77640-248 77660-584 77652-240
1-(3,4-Difluoro-2-methylphenyl)cyclobutanol ≥95%
1-(3,4-Difluoro-2-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD34774216
CAS Number: 2918873-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR