1-(3,4-Difluoro-2-methylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

94723-5G 94723-1G 94723-25G 94723-500MG
77630-926EA 1441 USD
77630-926 77640-246 77657-096 77648-230
1-(3,4-Difluoro-2-methylphenyl)cyclopentanol ≥95%
1-(3,4-Difluoro-2-methylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 212.236 g/mol
MW: C₁₂H₁₄F₂O
Storage Temperature: Ambient
MDL Number: MFCD34774215
CAS Number: 2918928-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR