1-(3-Fluoro-5-(trifluoromethyl)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

94703-5G 94703-25G 94703-1G 94703-500MG
77630-924EA 1441 USD
77630-924 77660-580 77644-228 77648-228
1-(3-Fluoro-5-(trifluoromethyl)phenyl)cyclopentanol ≥95%
1-(3-Fluoro-5-(trifluoromethyl)phenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 248.217 g/mol
MW: C₁₂H₁₂F₄O
Storage Temperature: Ambient
MDL Number: MFCD34774195
CAS Number: 2918946-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR