Ethyl 2-(5-chloro-2-fluoro-3-methylphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

94694-1G 94694-5G 94694-500MG
77640-242EA 647 USD
77640-242 77660-576 77652-236
Ethyl 2-(5-chloro-2-fluoro-3-methylphenyl)-2-oxoacetate ≥97%
Ethyl 2-(5-chloro-2-fluoro-3-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 244.647 g/mol
MW: C₁₁H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34774186
CAS Number: 1892301-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR