2-Fluoro-1-(2-fluoro-3-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

94676-5G 94676-500MG 94676-25G 94676-1G
77630-916EA 1441 USD
77630-916 77653-134 77660-574 77644-226
2-Fluoro-1-(2-fluoro-3-(trifluoromethyl)phenyl)ethanone ≥95%
2-Fluoro-1-(2-fluoro-3-(trifluoromethyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 224.127 g/mol
MW: C₉H₅F₅O
Storage Temperature: Ambient
MDL Number: MFCD34774171
CAS Number: 2918928-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR