1-(2,3-Dichloropyridin-4-yl)-2-fluoroethanone ≥97%

Supplier: AOBChem USA

Warning

94672-5G 94672-1G 94672-25G 94672-500MG
77630-914EA 1441 USD
77630-914 77640-236 77660-572 77652-232
1-(2,3-Dichloropyridin-4-yl)-2-fluoroethanone ≥97%
1-(2,3-Dichloropyridin-4-yl)-2-fluoroethanone

Precaución: For Research Use Only

Formula: 208.017 g/mol
MW: C₇H₄Cl₂FNO
Storage Temperature: Ambient
MDL Number: MFCD34774167
CAS Number: 2918847-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR