Ethyl 2-(2,6-dichloro-3-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

94655-5G 94655-25G 94655-1G 94655-500MG
77630-912EA 1441 USD
77630-912 77636-264 77644-224 77652-230
Ethyl 2-(2,6-dichloro-3-methoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,6-dichloro-3-methoxyphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 277.101 g/mol
MW: C₁₁H₁₀Cl₂O₄
Storage Temperature: Ambient
MDL Number: MFCD34774151
CAS Number: 2918946-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR