1-(3-Chloro-2-fluorophenyl)-2-fluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
94646-5G
94646-25G
94646-1G
94646-500MG
77630-908EA
1441
USD
77630-908
77660-570
77644-220
77648-220
1-(3-Chloro-2-fluorophenyl)-2-fluoroethanone ≥95%
1-(3-Chloro-2-fluorophenyl)-2-fluoroethanone
Precaución: For Research Use Only
Formula:
190.575 g/mol MW: C₈H₅ClF₂O Storage Temperature: Ambient |
MDL Number:
MFCD34774142 CAS Number: 2918873-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...