Ethyl 2-(2-bromo-5-fluoropyridin-4-yl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

94615-5G 94615-25G 94615-1G 94615-500MG
77630-906EA 1441 USD
77630-906 77636-262 77644-218 77648-218
Ethyl 2-(2-bromo-5-fluoropyridin-4-yl)-2-oxoacetate ≥97%
Ethyl 2-(2-bromo-5-fluoropyridin-4-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 276.059 g/mol
MW: C₉H₇BrFNO₃
Storage Temperature: Ambient
MDL Number: MFCD18910274
CAS Number: 2167849-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR