Ethyl 2-(2-chloro-3-fluoropyridin-4-yl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

94614-5G 94614-25G 94614-1G 94614-500MG
77630-904EA 1441 USD
77630-904 77636-260 77640-234 77648-216
Ethyl 2-(2-chloro-3-fluoropyridin-4-yl)-2-oxoacetate ≥95%
Ethyl 2-(2-chloro-3-fluoropyridin-4-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 231.608 g/mol
MW: C₉H₇ClFNO₃
Storage Temperature: Ambient
MDL Number: MFCD34774112
CAS Number: 2918919-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR