(4-(Trifluoromethyl)-1,2-phenylene)bis(methylsulfane) ≥95%

Supplier: AOBChem USA

Warning

94080-5G 94080-25G 94080-1G 94080-500MG
77633-864EA 1441 USD
77633-864 77660-568 77644-216 77652-228
(4-(Trifluoromethyl)-1,2-phenylene)bis(methylsulfane) ≥95%
(4-(Trifluoromethyl)-1,2-phenylene)bis(methylsulfane)

Precaución: For Research Use Only

Formula: 238.293 g/mol
MW: C₉H₉F₃S₂
Storage Temperature: Ambient
CAS Number: 3037695-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR