1-(Benzyloxy)-3-chloro-2,4-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

93550-5G 93550-1G 93550-25G 93550-500MG
77633-862EA 938 USD
77633-862 77640-232 77660-566 77648-214
1-(Benzyloxy)-3-chloro-2,4-difluorobenzene ≥95%
1-(Benzyloxy)-3-chloro-2,4-difluorobenzene

Precaución: For Research Use Only

Formula: C₁₃H₉ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD29044287
CAS Number: 1881289-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR