2-Chloro-4-ethoxy-1,3-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

93547-5G 93547-1G 93547-500MG 93547-25G
77633-860EA 1140 USD
77633-860 77640-230 77653-132 77660-564
2-Chloro-4-ethoxy-1,3-difluorobenzene ≥95%
2-Chloro-4-ethoxy-1,3-difluorobenzene

Precaución: For Research Use Only

Formula: C₈H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD28750536
CAS Number: 1805526-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR