2,4-Difluoro-3-chloroanisole ≥95%

Supplier: AOBChem USA

Warning

93546-5G 93546-1G 93546-25G 93546-500MG
77633-858EA 1029 USD
77633-858 77640-228 77657-090 77648-212
2,4-Difluoro-3-chloroanisole ≥95%
2,4-Difluoro-3-chloroanisole

Precaución: For Research Use Only

Formula: C₇H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD18415544
CAS Number: 1261472-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR