1-(5-Chloro-2-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

93467-5G 93467-1G 93467-25G 93467-500MG
77630-902EA 1441 USD
77630-902 77640-226 77660-562 77652-226
1-(5-Chloro-2-methylphenyl)cyclobutanol ≥95%
1-(5-Chloro-2-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD34773581
CAS Number: 2918947-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR