1-(4-Chloro-2-methoxyphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

93456-5G 93456-500MG 93456-25G 93456-1G
77630-900EA 1441 USD
77630-900 77653-130 77657-088 77644-214
1-(4-Chloro-2-methoxyphenyl)cyclopentanol ≥95%
1-(4-Chloro-2-methoxyphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 226.699 g/mol
MW: C₁₂H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34773570
CAS Number: 2918906-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR