1-(4-Chloro-2-methoxyphenyl)-2-fluoroethanone ≥97%

Supplier: AOBChem USA

Warning

93455-1G 93455-5G 93455-500MG
77635-370EA 967 USD
77635-370 77657-086 77648-210
1-(4-Chloro-2-methoxyphenyl)-2-fluoroethanone ≥97%
1-(4-Chloro-2-methoxyphenyl)-2-fluoroethanone

Precaución: For Research Use Only

Formula: 202.61 g/mol
MW: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD24539391
CAS Number: 2385174-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR