1-(2-Chloro-4-methoxyphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

93437-5G 93437-1G 93437-25G 93437-500MG
77633-856EA 1357 USD
77633-856 77640-224 77657-084 77648-208
1-(2-Chloro-4-methoxyphenyl)cyclopentanol ≥95%
1-(2-Chloro-4-methoxyphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 226.699 g/mol
MW: C₁₂H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34773552
CAS Number: 2918947-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR