1-(3-Methoxy-5-(trifluoromethyl)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

93427-5G 93427-1G 93427-25G 93427-500MG
77630-896EA 1441 USD
77630-896 77640-220 77657-080 77652-224
1-(3-Methoxy-5-(trifluoromethyl)phenyl)cyclobutanol ≥95%
1-(3-Methoxy-5-(trifluoromethyl)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 246.226 g/mol
MW: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34773543
CAS Number: 2918918-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR