1-(5-Chloro-2-methoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

93425-1G 93425-5G 93425-500MG
77640-218EA 531 USD
77640-218 77657-078 77652-222
1-(5-Chloro-2-methoxyphenyl)cyclobutanol ≥95%
1-(5-Chloro-2-methoxyphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD21788666
CAS Number: 1537951-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR