1-(3-Fluoro-2-methoxyphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

93421-5G 93421-1G 93421-500MG
77630-894EA 1658 USD
77630-894 77640-216 77652-220
1-(3-Fluoro-2-methoxyphenyl)cyclopentanol ≥95%
1-(3-Fluoro-2-methoxyphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 210.245 g/mol
MW: C₁₂H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD34773538
CAS Number: 2918846-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR