1-(3-Chloro-2-methoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

93405-5G 93405-1G 93405-25G 93405-500MG
77630-890EA 1441 USD
77630-890 77640-212 77660-558 77648-204
1-(3-Chloro-2-methoxyphenyl)cyclobutanol ≥95%
1-(3-Chloro-2-methoxyphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34773524
CAS Number: 2918830-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR