1-(3-Chloro-5-methoxyphenyl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

93399-5G 93399-1G 93399-500MG
77657-076EA 1811 USD
77657-076 77644-212 77652-216
1-(3-Chloro-5-methoxyphenyl)cyclobutanol ≥97%
1-(3-Chloro-5-methoxyphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34773518
CAS Number: 2021180-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR