1-(3-Fluoro-5-methoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

93394-5G 93394-1G 93394-25G 93394-500MG
77630-888EA 1441 USD
77630-888 77640-210 77657-074 77648-202
1-(3-Fluoro-5-methoxyphenyl)cyclobutanol ≥95%
1-(3-Fluoro-5-methoxyphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34773513
CAS Number: 1934445-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR