Ethyl 2-(4-(methoxymethoxy)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

93380-5G 93380-25G 93380-1G 93380-500MG
77630-886EA 1441 USD
77630-886 77636-254 77640-206 77648-200
Ethyl 2-(4-(methoxymethoxy)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-(methoxymethoxy)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 238.237 g/mol
MW: C₁₂H₁₄O₅
Storage Temperature: Ambient
MDL Number: MFCD34818534
CAS Number: 2969366-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR